Learn More
Strem, An Ascensus Company CAS 654057-97-3. 10g. Trihexyl(tetradecyl)phosphonium bromide min. 95% CYPHOS IL 102. MFCD04117697
SDP
Supplier:  Strem, An Ascensus Company 15637810G
Encompass
CAS 654057-97-3. 10g. Trihexyl(tetradecyl)phosphonium bromide min. 95% CYPHOS IL 102. MFCD04117697. Molecular Weight 563.76. Molecular Formula [(C6H13)3(C14H29)P]Brae. Color/form pale yellow liq. Strem 15-6378. http//www.strem.com/catalog/v/15-6378/
Catalog No. 50-211-0770