Learn More
Medchemexpress LLC Cyproconazole | 94361-06-5 | 98.7% | C15H18ClN3O | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA0277R50MG
Encompass_Preferred
Cyproconazole is a triazole fungicide that effectively protects crops, fruits, and vegetables from a wide range of fungal pathogens. It has been observed to induce hepatocellular adenomas and carcinomas in CD-1 mice. Furthermore, this compound demonstrates low toxicity to zebrafish embryos and impacts their locomotor activity.
  • Triazole fungicide
  • Protects against fungal pathogens
  • Low toxicity to zebrafish embryos
  • Affects zebrafish locomotor activity
Catalog No. 50-004-32951