Learn More
Medchemexpress LLC Cyprodinil 10mM 1mL | 121552-61-2 | 225.29 g/mol | C14H15N3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11621410MM 1ML
Encompass_Preferred
Cyprodinil is an anilinopyrimidine broad-spectrum fungicide supplied as a research-grade reagent. The product is provided as a ready-to-use 10 mM solution in DMSO (1 mL) for laboratory and analytical applications; solid quantities are also available.
  • Ready-to-use 10 mM solution in DMSO.
  • Packaged as a 1 mL vial.
  • High reported purity (98.84%).
  • Suitable for analytical and research applications.
  • Includes CAS identifier (121552-61-2) for traceability.
Catalog No. 50-002-27306