Learn More
Medchemexpress LLC Cyprodinil Standard 50mg | 121552-61-2 | 225.29 g/mol | C14H15N3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY116214R50MG
Encompass_Preferred
Cyprodinil (Standard) is an analytical reference standard of cyprodinil (CAS 121552-61-2) supplied as a 50 mg vial for research and analytical use. It is intended for calibration, method development, and quality control testing in residue and environmental analysis.
  • High purity suitable for analytical applications (99.04%).
  • Compact 50 mg package for convenient handling and storage.
  • Molecular formula C14H15N3 and molecular weight 225.29 g/mol.
  • Suitable for calibration, method development, and QC testing workflows.
  • Stable solid standard for use in HPLC, GC, and mass spectrometry methods.
Catalog No. 50-002-29487