Learn More
Cayman Chemical Cyproteron Acetate 250mg
SDP
Supplier:  Cayman Chemical 16622250 MG
Encompass_Preferred
An androgen receptor antagonist; binds to human androgen receptors (Ki = 14 nM); inhibits dihydrotestosterone-induced androgen receptor activation in CV-1 cells (IC50 = 26 nM); decreases ventral prostate weight in castrated immature rats at 30 mg/kg; suppresses accessory sexual glands and fertility in adult male rats at 10 mg/kg; induces apoptosis in primary adult female rat hepatocytes at 0.3 µM
Catalog No. 50-296-7499