Learn More
Medchemexpress LLC Cyproterone | 2098-66-0 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1623
Encompass_Preferred
Cyproterone is an antiandrogen that suppresses the actions of testosterone by blocking androgen receptors. Its acetate form can be used in the research of hypersexuality and prostate cancer.
  • Antiandrogen
  • Suppresses testosterone actions
  • Blocks androgen receptors
  • Acetate form used in research for hypersexuality and prostate cancer
Catalog No. 50-004-14137