Learn More
Medchemexpress LLC Cyproterone | 2098-66-0 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB162350MG
Encompass_Preferred
Cyproterone is an antiandrogen that suppresses the actions of testosterone by blocking androgen receptors. Its acetate form can be used in the research of hypersexuality and prostate cancer. This product is intended for research use only and not for patient use.
  • Functions by blocking androgen receptors
  • Suppresses the actions of testosterone
  • Acetate form used in hypersexuality and prostate cancer research
  • Intended for research use only
Catalog No. 50-004-13264