Learn More
Medchemexpress LLC Cyproterone | 2098-66-0 | MFCD00869226 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB16235MG
Encompass_Preferred
Cyproterone is an antiandrogen that suppresses the actions of testosterone via blocking androgen receptors. Cyproterone's acetate form can be used in the research of hypersexuality and prostate cancer.
Features and benefits
  • Suppresses the actions of testosterone.
  • Functions by blocking androgen receptors.
  • Its acetate form can be used in the research of hypersexuality.
  • Its acetate form can be used in the research of prostate cancer.
  • Has been investigated in clinical trials for conditions such as acne vulgaris.
  • Has been investigated in clinical trials for polycystic ovary syndrome.
  • Has been investigated in clinical trials for gender dysphoria.
  • Has been investigated in clinical trials for hirsutism, androgenetic alopecia, acne and seborrhea in women.
  • Has been investigated in clinical trials for hyperandrogenism.
Catalog No. 50-004-13089