Learn More
Selleck Chemical LLC Cyproterone Acetate S2042-100mg
SDP
Supplier:  Selleck Chemical LLC S2042100MG
Encompass_Preferred
Cyproterone acetate is an androgen receptor (AR) antagonist with IC50 of 7 1 nM as well as a weak progesterone receptor agonist with weak pro-gestational and glucocorticoid activity
Catalog No. 50-311-0350