Learn More
Medchemexpress LLC Cyromazine (Standard) | 66215-27-8 | 99.7% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1331R50MG
Encompass_Preferred
Cyromazine (Standard) is an analytical standard of Cyromazine, intended for research and analytical applications. It is a triazine insect growth regulator, cyclopropyl derivative of melamine, used as an insecticide and an acaricide. It affects the nervous system of immature larval stages of certain insects and interferes with the Ecdysone signaling pathway.
  • Analytical standard grade for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Insect growth regulator.
  • Cyclopropyl derivative of melamine.
  • Affects the nervous system of immature larval stages of certain insects.
  • Used as an insecticide and an acaricide.
Catalog No. 50-004-13239