Learn More
Medchemexpress LLC Cyromazine (Standard) | 66215-27-8 | 99.71% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB1331R
Encompass_Preferred
Cyromazine (Standard) is the analytical standard of Cyromazine, intended for research and analytical applications. It is a triazine insect growth regulator used as an insecticide and acaricide, functioning by affecting the nervous system of immature larval stages of certain insects. The compound is an analytical standard used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Used as an insecticide and acaricide
  • Functions by affecting the nervous system of immature larval stages
  • Reduces germ cells by interfering with the Ecdysone signaling pathway
  • Induces renal toxicity, which can be attenuated by green tea extract
  • Applicable in HPLC, GC, and MS research experiments
Catalog No. 50-004-14410