Learn More
Molecular Dimensions CYROMOUNT SET .1MM
SDP
Supplier:  Molecular Dimensions MD7134CM
Encompass_Preferred
Protein Crystallography. A set of mounted 0.10 mm LithoLoops mounted in CryoCaps and Magnetic CryoVials. Each CryoCap is DataMatrix etched with the size of the LithoLoop. The sets are provided in a handy storage box.
Catalog No. NC0071661