Learn More
Origen Biomedical CryoStore 250 EVA Freezing Bag, 30 – 70ml, “N” Tube Set, double aseptically packed | 12/case
Supplier:  Origen Biomedical CS250NS
Encompass
The CryoStore™ bag is designed to freeze blood components. One male luer with cap, two female luers with caps, and a needle-free injection port.
Catalog No. NC1741452