Learn More
Medchemexpress LLC Cys-McMMAF | 1160590-05-5 | 99.5% | 1046.32 g/mol | C52H83N7O13S | 1 MG

Supplier: Medchemexpress LLC HY1502331MG
Cys-McMMAF is the cysteine-linked MMAF payload released from an antibody-drug conjugate. It is an MMAF derivative joined via a maleimidocaproyl linker, supplied as a high-purity research reagent for use in preclinical antibody-drug conjugate studies.
- High purity 99.5%.
- Molecular weight 1046.32 g/mol and formula C52H83N7O13S.
- Cas number 1160590-05-5.
- Supplied in small milligram quantities suitable for preclinical research.
- Intended for use as a cysteine-linked payload in ADC payload characterization and in vitro or in vivo studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.