Learn More
Cayman Chemical CysLT2 ReceptrN-Term Poly 1ea
SDP
Supplier:  Cayman Chemical 1205601 EA
Encompass_Preferred
Immunogen: Peptide corresponding to the N-terminal region of human CysLT2 • Host: Rabbit • Species Reactivity: (+) Human; (-) Rat • Cross Reactivity: (+) CysLT2; (-) CysLT1 • Applications: ELISA, FC, WB
Catalog No. 50-295-4378