Learn More
Cayman Chemical Cysmethynil 10MG
SDP
Supplier:  Cayman Chemical 1474510 MG
Encompass_Preferred
An indole-based, time-dependent inhibitor of Icmt (IC50 = i = 0.14 µM for the final complex); decreases mTOR signaling, accumulation of cells in the G1 phase, and autophagy-mediated cell death in PC3 prostate cancer cells
Catalog No. 50-297-2905