Learn More
Selleck Chemical LLC Cystamine dihydrochloride S3695-1g
SDP
Supplier:  Selleck Chemical LLC S36951G
Encompass_Preferred
Cystamine dihydrochloride (Decarboxycystine 2-Aminoethyl disulfide 2 2 -Dithiobisethanamine) acts as an anti-infective agent which is used in the treatment of urinary tract infections and also as a radiation-protective agent that interferes with sulfhydryl enzymes
Catalog No. 50-310-8949