Learn More
Cayman Chemical CystathIonIn-d4 500ug
SDP
Supplier:  Cayman Chemical 36610500 UG
Encompass_Preferred
An internal standard for the quantification of cystathionine by GC- or LC-MS
Catalog No. 50-290-9289