Learn More
Genetex Inc Cystatin A Monoclonal Antibody [MSVA-461M] HistoMAX™ (GTX04453) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc CYSTATIN A ANTIBDY MSVA4
Encompass_Preferred
The Cystatin A antibody [MSVA-461M] HistoMAX™ (GTX04453) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human skin tissue using GTX04453 Cystatin A antibody [MSVA-461M] HistoMAX. In the skin a nuclear and cytoplasmic Cystatin A immunostaining is predominantly seen in the granular cell layer. The staining intensity decreases towards the basal cell layer.
Catalog No. NC3667726