Learn More
Medchemexpress LLC Cysteine Protease inhibitor (hydrochloride) | 2197053-49-7 | 96.0% | 338.79 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17541A
Encompass_Preferred
Cysteine Protease inhibitor hydrochloride is a cysteine protease inhibitor that can be used in immunoblotting experiments of cells or tissues.
  • Cysteine protease inhibitor.
  • Can be used in immunoblotting experiments of cells or tissues.
Catalog No. 50-003-84358