Learn More
Selleck Chemical LLC Momelotinib (CYT387) 50mg 1056634-68-4 LM-1149 , CYT11387
SDP
Supplier:  Selleck Chemical LLC S221950MG
Encompass_Preferred
Momelotinib (CYT387, LM-1149 , CYT11387) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50 of 11 nM/18 nM, ~10-fold selectivity versus JAK3. Momelotinib (CYT387) induces apoptosis and autophagy. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-877-6