Learn More
Cayman Chemical CytarabIn 5-monphosphate 10mg
SDP
Supplier:  Cayman Chemical 3649610 MG
Encompass_Preferred
An active metabolite of cytarabine; is incorporated into DNA by DNA polymerase α, which slows the rate of DNA synthesis by approximately 2000-fold in a cell-free assay; inhibits nuclear and mitochondrial DNA replication in S. cerevisiae at 15 mM; improves survival in an L1210 murine leukemia model at 3.5-75.1 mg/kg
Catalog No. 50-291-2309