Learn More
Medchemexpress LLC Cytarabine-d2 | 40632-26-6 | 98.4% | 245.23 g/mol | C9H11D2N3O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13605S5MG
Encompass_Preferred
Cytarabine-d2 is the deuterium-labeled stable isotope of cytarabine provided as an analytical reference standard for research use. As a nucleoside analog it causes S-phase cell cycle arrest and inhibits DNA polymerase; the deuterated form is intended for use as an internal standard in mass spectrometry and isotope tracing studies.
  • Stable isotope-labeled internal standard for mass spectrometry.
  • High purity (98.4%) suitable for analytical use.
  • Available in small research pack sizes such as 5 mg.
  • CAS number 40632-26-6 and formula C9H11D2N3O5.
  • Commonly used in drug metabolism and tracer studies.
Catalog No. 50-002-22200