Learn More
Cayman Chemical CytIdIn 3-monphosphate 10mg
SDP
Supplier:  Cayman Chemical 3383310 MG
Encompass_Preferred
A ribonucleotide; produced by hydrolysis of cytidine 2’,3’-cyclic monophosphate via RNase; inhibits RNase; can be dephosphorylated to cytidine by 3’-nucleotidase
Catalog No. 50-294-6609