Learn More
Cayman Chemical CytIdIn-d2 500ug
SDP
Supplier:  Cayman Chemical 36320500 UG
Encompass_Preferred
An internal standard for the quantification of cytidine by GC- or LC-MS
Catalog No. 50-291-2593