Learn More
Medchemexpress LLC Cytidine-13C | 201996-57-8 | 99.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0158S150MG
Encompass_Preferred
Cytidine-13C is the 13C labeled Cytidine. Cytidine is a pyrimidine nucleoside and acts as a component of RNA. It is a precursor of uridine. Cytidine controls neuronal-glial glutamate cycling, affecting cerebral phospholipid metabolism and catecholamine. For research use only. Not for sale to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06260