Learn More
Medchemexpress LLC Cytidine-13C | 201996-57-8 | MFCD00209706 | >99.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0158S15MG
Encompass_Preferred
Cytidine-13C is the 13C labeled Cytidine. It is a pyrimidine nucleoside and acts as a component of RNA. Cytidine is a precursor of uridine and controls neuronal-glial glutamate cycling, affecting cerebral phospholipid metabolism, and catecholamine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Labeled with 13C for isotopic studies
  • Functions as a pyrimidine nucleoside
  • Essential component of RNA
  • Precursor to uridine
  • Regulates neuronal-glial glutamate cycling
  • Affects cerebral phospholipid metabolism
  • Influences catecholamine levels
  • Suitable for use as a tracer in analytical techniques
  • Can be used as an internal standard for quantitative analysis
Catalog No. 50-004-06943