Learn More
Medchemexpress LLC Cytidine, 2'-deoxy-2',2'-difluoro-, hydrochloride | 122111-03-9 | 99.9% | C9H12ClF2N3O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB000310MM/1ML
Encompass_Preferred
Gemcitabine hydrochloride is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. It inhibits DNA synthesis and repair, leading to autophagy and apoptosis.
  • Targets DNA/RNA synthesis
  • Functions as a nucleoside antimetabolite/analog
  • Induces autophagy and apoptosis
  • Used in cancer research
Catalog No. 50-004-32295