Learn More
Medchemexpress LLC Cytidine 5'-diphosphate (trisodium salt) | 34393-59-4 | 98.0% | 469.12 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW008915
Encompass_Preferred
Cytidine 5'-diphosphate trisodium salt (CDP) is generated through the transfer of a phosphoryl group from ATP to cytidine monophosphate (CMP), a reaction catalyzed by uridine monophosphate kinase (UMPK). This compound can be utilized to synthesize Cytidine triphosphate (CTP), which is essential for the creation of DNA and RNA.
  • Produced by the transfer of phosphoryl group from ATP to cytidine monophosphate (CMP) catalyzed by uridine monophosphate kinase (UMPK).
  • Can be used to produce Cytidine triphosphate (CTP) for synthesis of DNA and RNA.
  • CDP-ribitol can be used for the diagnostics and research of CDP-l-ribitol pyrophosphorylase A (CRPPA) muscular dystrophy.
  • For research use only.
Catalog No. 50-003-66696