Learn More
Selleck Chemical LLC Cytisine 25mg 485-35-8 Baphitoxine,Sophorine
SDP
Supplier:  Selleck Chemical LLC S228725MG
Encompass_Preferred
Cytisine (Baphitoxine, Sophorine) is a nicotinic acetylcholine receptor agonist. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-521-4