Learn More
Genetex Inc Cytokeratin 5 Monoclonal Antibody [MSVA-605M] HistoMAX™ (GTX04402) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04402
Encompass_Preferred
The Cytokeratin 5 antibody [MSVA-605M] HistoMAX™ (GTX04402) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human squamous cell carcinoma (SCC) from lung tissue using GTX04402 Cytokeratin 5 antibody [MSVA-605M] HistoMAX. Strong cytokeratin 5 immunostaining in a squamous cell carcinoma of the lung..
Catalog No. NC3666769