Learn More
Genetex Inc Cytokeratin 10 Monoclonal Antibody [MSVA-610M] HistoMAX™ (GTX04401) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04401
Encompass_Preferred
The Cytokeratin 10 antibody [MSVA-610M] HistoMAX™ (GTX04401) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human esophagus tissue using GTX04401 Cytokeratin 10 antibody [MSVA-610M] HistoMAX. Absence of cytokeratin 10 staining in non keratinizing squamous epithelium of the esophagus.
Catalog No. NC3666767