Learn More
Genetex Inc Cytokeratin 13 Monoclonal Antibody [MSVA-613M] HistoMAX™ (GTX04385) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04385
Encompass_Preferred
The Cytokeratin 13 antibody [MSVA-613M] HistoMAX™ (GTX04385) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human skin tissue using GTX04385 Cytokeratin 13 antibody [MSVA-613M] HistoMAX. Absence of cytokeratin 13 staining in the keratinizing squamous epithelium of the skin.
Catalog No. NC3666316