Learn More
Genetex Inc Cytokeratin 14 Monoclonal Antibody [MSVA-614R] HistoMAX™ (GTX04377) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04377
Encompass_Preferred
The Cytokeratin 14 antibody [MSVA-614R] HistoMAX™ (GTX04377) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human esophageal squamous cell carcinoma (ESCC) tissue using GTX04377 Cytokeratin 14 antibody [MSVA-614R] HistoMAX. Squamous cell carcinoma of the esophagus with strong predominantly basal cytokeratin 14 immunostaining.
Catalog No. NC3665628