Learn More
Genetex Inc Cytokeratin 15 Monoclonal Antibody [MSVA-615M] HistoMAX™ (GTX04411) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04411
Encompass_Preferred
The Cytokeratin 15 antibody [MSVA-615M] HistoMAX™ (GTX04411) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human esophagus tissue using GTX04411 Cytokeratin 15 antibody [MSVA-615M] HistoMAX. A strong cytokeratin 15 immunostaining is always found in all cell layers of all non keratinizing and keratinizing squamous epithelium.
Catalog No. NC3666784