Learn More
Genetex Inc Cytokeratin 18 Monoclonal Antibody [MSVA-118R] HistoMAX™ (GTX04374) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04374
Encompass_Preferred
The Cytokeratin 18 antibody [MSVA-118R] HistoMAX™ (GTX04374) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human gastric antrum tissue using GTX04374 Cytokeratin 18 antibody [MSVA-118R] HistoMAX. Strong cytokeratin 18 staining of all epithelial cells.
Catalog No. NC3665624