Learn More
Genetex Inc Cytokeratin 19 Monoclonal Antibody [MSVA-119M] HistoMAX™ (GTX04378) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04378
Encompass_Preferred
The Cytokeratin 19 antibody [MSVA-119M] HistoMAX™ (GTX04378) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human malignant mesothelioma tissue using GTX04378 Cytokeratin 19 antibody [MSVA-119M] HistoMAX. Malignant mesothelioma with strong cytokeratin 19 positivity.
Catalog No. NC3666282