Learn More
Aat Bioquest CYTO-13 5 mM DMSO solution (0.5 mL)
SDP
Supplier:  Aat Bioquest 17553
Encompass_Preferred
CYTO-13 is a fluorogenic DNA-selective and cell-permeant dye for analyzing DNA content in living cells. This catalog is for 0.5 mL.
Catalog No. 50-224-9744