Learn More
Cayman Chemical CytochalasIn J 5mg
SDP
Supplier:  Cayman Chemical 184435 MG
Encompass_Preferred
A deacetylated analog of cytochalasin H that weakly inhibits actin assembly but significantly alters mitotic spindle microtubule organization and kinetochore structure; suppresses the production of reactive oxygen species by stimulated human neutrophils
Catalog No. 50-296-5040