Learn More
CPC Scientific H-Ala-Asn-Glu-Arg-Ala-Asp-Leu-Ile-Ala-Tyr-Leu-Lys-Gln-Ala-Gly-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CYTC001A
Encompass
This peptide is an antigenic peptide that prevents the development of mature 2B4 TCRhi CD4+8+ thymocytes, competitively blocks antigen induced IL-2 production plays major role in T-cell selection.ONE-LETTER SEQUENCE: ANERADLIAYLKQAGKMOLECULAR FORMULA: C77H129N23O24MOLECULAR WEIGHT:1761.01STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [213260-63-0], RESEARCH AREA: ImmunomodulationREFERENCES: R.Wang et al., Proc. Natl. Acad. Sci. USA, 95, 3804 (1998)
Catalog No. 50-210-9177