Learn More
Apexbio Technology LLC Cytochrome c - pigeon (88-104) 86579-06-8 1mg
SDP
Supplier:  Apexbio Technology LLC A10611
Encompass_Preferred
Cytochrome c - pigeon 88-104 is a synthetic peptide recognized by pigeon cytochrome c-specific T cells via presentation on the mouse MHC class II molecule I-E It is designed to serve as an antigenic model to investigate T cell-MHC interactions and antigen processing Cytochrome c - pigeon 88-104 exerts its biological activity primarily by activating antigen-specific T cell responses through MHC class II-restricted presentation In immunological studies activation is observed in T cells specific for pigeon cytochrome c when presented by I-E Based on these attributes Cytochrome c - pigeon 88-104 is a valuable tool for research in T cell recognition antigen presentation and mechanisms of adaptive immunity
Catalog No. 50-000-96451