Learn More
Medchemexpress LLC Cytokeratin 19 Antibody (PBS only) | 100 UG

Supplier: Medchemexpress LLC HYP81183A100UG
The Cytokeratin 19 Antibody (PBS only) is a rabbit-derived, non-conjugated IgG polyclonal antibody specifically targeting Cytokeratin 19. Supplied as a lyophilized powder in PBS, this affinity-purified antibody is involved in the organization of myofibers and helps link the contractile apparatus to dystrophin at costameres of striated muscle.
- Rabbit-derived, non-conjugated IgG polyclonal antibody
- Targets Cytokeratin 19
- Reactivity: Human, Mouse
- Observed molecular weight: 42 kDa
- Formulated in PBS as a lyophilized powder
- Stored at 2-8°C for 1 year; protect from light; do not freeze
- Applications include WB (1:500-2000), ELISA (1:5000-10000), IHC-P (1:100-500), IHC-F (1:100-500), FC (1μg/test), and ICC/IF (1:100-500)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.