Learn More
Medchemexpress LLC Cytokeratin 19 Antibody (PBS only) | 200 UG

Supplier: Medchemexpress LLC HYP81183A200UG
Cytokeratin 19 Antibody (PBS only) is a rabbit-derived, non-conjugated IgG polyclonal antibody that targets Cytokeratin 19. Cytokeratin is a protein crucial for the organization of myofibers, and along with KRT8, it helps connect the contractile apparatus to dystrophin at the costameres of striated muscle.
- Rabbit host
- Reactivity: Human, Mouse
- Non-conjugated
- Supplied in PBS
- Storage at 2-8°C for 1 year, protected from light, do not freeze
- Lyophilized powder appearance
- Shipped with blue ice
- Applications: WB, ELISA, IHC-P, IHC-F, FC, ICC/IF
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.