Learn More
Medchemexpress LLC Cytokeratin 19 Antibody (PBS only) | 50 UG
SDP
Supplier:  Medchemexpress LLC HYP81183A50UG
Encompass_Preferred
Cytokeratin 19 Antibody (PBS only) is a rabbit-derived, non-conjugated IgG polyclonal antibody designed to target Cytokeratin 19. Cytokeratin is a protein involved in the organization of myofibers, helping to link the contractile apparatus to dystrophin at the costameres of striated muscle.
  • Rabbit-derived polyclonal antibody
  • Targets Cytokeratin 19
  • Non-conjugated IgG
  • Suitable for WB, ELISA, IHC-P, IHC-F, FC, and ICC/IF applications
  • Observed band size: 42 kDa
Catalog No. 50-004-42445