Learn More
Medchemexpress LLC Cytokeratin 4 Antibody (YA1762) | 50 UL

Supplier: Medchemexpress LLC HYP8201750UL
The Cytokeratin 4 Antibody (YA1762) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Cytokeratin 4. Cytokeratin 4 (KRT4) is a type II cytokeratin found in the differentiated layers of mucosal and esophageal epithelium, typically paired with cytokeratin 13.
- Host: rabbit
- Reactivity: human, rat
- Isotype: IgG
- Conjugation: non-conjugated
- Purity: affinity purified
- Formulation: supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide, and 0.05% BSA
- Storage: stored at -20°C for 1 year
- Avoid repeated freeze/thaw cycles
- Applications: WB (Western Blot), IP (Immunoprecipitation)
- Predicted molecular weight: 56 kDa
- Observed molecular weight: 56 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.