Learn More
Medchemexpress LLC Cytokeratin 5 antibody (YA3786)(PBS only) | 62 kDa | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8408910UL
Encompass_Preferred
Cytokeratin 5 Antibody (YA3786) is a mouse-derived, non-conjugated IgG1 monoclonal antibody targeting Cytokeratin 5. It is supplied in PBS, pH 7.4, and is suitable for various applications including WB, IHC-P, ICC/IF, FC, and ELISA. This antibody is intended for research use only.
  • Mouse-derived IgG1 monoclonal antibody
  • Targets Cytokeratin 5
  • Non-conjugated
  • Suitable for Western Blot, Immunohistochemistry-Paraffin, Immunocytochemistry/Immunofluorescence, Flow Cytometry, and ELISA
  • Stored at -20°C for 1 year
Catalog No. 50-004-48386