Learn More
Cayman Chemical CytokRatIn 18 RabbIt Monclo
SDP
Supplier:  Cayman Chemical 32229100 UL
Encompass_Preferred
A fungal metabolite with diverse biological activities cytotoxic to BC-1 human breast cancer cells (IC50 5 Ug/ml) active against L. braziliensis amastigotes (IC50 78.5 UM) active against the plant pathogenic fungi C. gloeosporioides (MIC 292 UM)
Catalog No. 50-294-7381