Learn More
Bioworld CYTOPLASMIC & NUCLEAR RNA P
Supplier:  Bioworld 107600631
Encompass_Preferred
CYTOPLASMIC & NUCLEAR RNA PURIFICATION KIT: For the convenient purification of cytoplasmic and nuclear RNA from cultured cells and tissues High quality RNA; No need for phenol; Genomic DNA-free cytoplasmic RNA. FEATURES AND BENEFITS Fas...
Catalog No. 50-196-917