Learn More
Medchemexpress LLC Cytosine-13C,15N2 | 181517-10-2 | 98.0% | 113.07 g/mol | C3[13C]H5N[15N]2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYI0626S110MG
Encompass_Preferred
Cytosine-13C,15N2 is a stable isotope-labeled cytosine enriched with carbon-13 and nitrogen-15, supplied as a white to off-white solid for use as a tracer or internal standard in analytical methods such as NMR, GC-MS, and LC-MS. Intended for research use only; store at 4°C and protect from light; in solution store at -80°C up to 6 months or -20°C up to 1 month (protect from light).
  • Stable isotope labeling with 13C and 15N for tracing and quantitation.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High purity (≥98%).
  • Available in small packaging sizes for analytical workflows.
  • White to off-white solid; protect from light during storage.
Catalog No. 50-002-97168