Learn More
Medchemexpress LLC Cytosine-13C,15N2 | 181517-10-2 | 98% | 113.07 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYI0626S11MG
Encompass_Preferred
Cytosine-13C,15N2 is the 13C and 15N labeled Cytosine. Cytosine is one of the four main bases found in DNA and RNA. Cytosine modifications exhibit circadian oscillations that are involved in epigenetic diversity and aging. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules as tracers
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs
  • Appears as a white to off-white solid
  • Should be stored at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protected from light)
Catalog No. 50-004-28749